335959
Bismuth(III) oxide
99.99%
Synonym(s):
Dibismuth trioxide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
99.99%
SMILES string
O=[Bi]O[Bi]=O
InChI
1S/2Bi.3O
InChI key
WMWLMWRWZQELOS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Preparation of new stabilized, oxide ion-conducting, bismuth vanadate phases by a microwave assisted method, from V2O5, Bi2O3 and other solid oxides, was reported. These ceramics show promise in solid oxide fuel cells, water-vapor electrolyzers and oxygen sensors.
Features and Benefits
Preparation of new stabilized, oxide ion-conducting, bismuth vanadate phases by a microwave assisted method, from V2O5, Bi2O3 and other solid oxides, was reported. These ceramics show promise in solid oxide fuel cells, water-vapor electrolyzers and oxygen sensors.
replaced by
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 83(1), 94-99 (2011-09-06)
Glasses with composition 20CdO·xBi(2)O(3)·(79.5-x)B(2)O(3) (15≤x≤35, x in mol%) containing 0.5 mol% of Er(3+) ions were prepared by melt-quench technique (1150°C in air). The amorphous nature of the glasses was confirmed by X-ray diffraction. The spectroscopic properties of the glasses were
Journal of the Air & Waste Management Association (1995), 62(9), 1032-1039 (2012-10-02)
Photocatalytic oxidation (PCO) is a promising technology for indoor air purification due to low operating cost, potentially long service life, and low maintenance. Ultraviolet light-emitting diode (UVLED) is a new concept in the field of PCO, which has several advantages
Chemistry of Materials, 10, 3400-3400 (1998)
Acta crystallographica. Section A, Foundations of crystallography, 68(Pt 1), 148-155 (2011-12-22)
X-ray diffraction from films consisting of layers with different thicknesses, structures and chemical contents is analysed. The disorder is described by probabilities for different sequences of layers. Closed analytical expressions for the diffracted X-ray intensity are obtained when the layers
Journal of endodontics, 38(3), 324-329 (2012-02-22)
Mineral trioxide aggregate (MTA) and Portland cement have been shown to be bioactive because of their ability to produce biologically compatible carbonated apatite. This study analyzed the interaction of MTA and white Portland cement with dentin in vivo. Seventy-two human dentin
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service