Skip to Content
Merck
All Photos(2)

Key Documents

298123

Sigma-Aldrich

Samarium(III) nitrate hexahydrate

99.9% trace metals basis

Synonym(s):

Samarium trinitrate, Trinitric acid samarium(III)·6hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sm(NO3)3 · 6H2O
CAS Number:
Molecular Weight:
444.47
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.9% trace metals basis
form:
powder and chunks

Quality Level

Assay

99.9% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: samarium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

SMILES string

[Sm+3].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/3NO3.6H2O.Sm/c3*2-1(3)4;;;;;;;/h;;;6*1H2;/q3*-1;;;;;;;+3

InChI key

HDCOFJGRHQAIPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Samarium(III) nitrate hexahydrate is a Lewis acid catalyst that is used in the synthesis of a variety of materials for applications such as:
  • synthesis of nitrate precursor solution which can be used as a nanocatalyst in the solid oxide regenerative fuel cells.
  • for the formation of coumarin derivative by von Pechmann condensation of phenol with ethyl acetoacetate in the presence of samarium complex as catalyst.
  • for the preparation of samarium doped ceria, which can be used in the fabrication of electrolyte for fuel cells.

Other Notes

Hydration may vary slightly

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of nanocrystalline samarium-doped CeO2 (SDC) powders as a solid electrolyte by using a simple solvothermal route
Karaca T, et al.
Ceramics International, 36(3), 1101-1107 (2010)
High performance air electrode for solid oxide regenerative fuel cells fabricated by infiltration of nano-catalysts
Lee S, et al.
Journal of Power Sources, 250, 15-20 (2014)
Samarium (III) catalyzed one-pot construction of coumarins
Bahekar SS and Shinde DB
Tetrahedron Letters, 45(43), 7999-8001 (2004)

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service