Skip to Content
Merck
All Photos(1)

Key Documents

335819

Sigma-Aldrich

Ethyl 3-bromobenzoate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€246.00

€246.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€246.00

About This Item

Linear Formula:
BrC6H4CO2C2H5
CAS Number:
Molecular Weight:
229.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€246.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.541 (lit.)

bp

130-131 °C/12 mmHg (lit.)

density

1.431 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)c1cccc(Br)c1

InChI

1S/C9H9BrO2/c1-2-12-9(11)7-4-3-5-8(10)6-7/h3-6H,2H2,1H3

InChI key

QAUASTLEZAPQTB-UHFFFAOYSA-N

Application

Ethyl 3-bromobenzoate was used in the preparation of ethyl 3-ethynylbenzoate.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Iijima et al.
Chemical & pharmaceutical bulletin, 47(3), 398-404 (1999-04-23)
Synthesis and biological evaluation of the first dicarba-closo-dodecaborane (carborane) derivatives of retinoids are described. Their retinoidal activity were examined in terms of the differentiation-inducing ability toward human promyelocytic leukemia HL-60 cells. High retinoidal activity (agonist or antagonist for retinoic acid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service