Skip to Content
Merck
All Photos(1)

Key Documents

234621

Sigma-Aldrich

Methyl 3-chlorobenzoate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€38.70
25 G
€96.10

€38.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€38.70
25 G
€96.10

About This Item

Linear Formula:
ClC6H4CO2CH3
CAS Number:
Molecular Weight:
170.59
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€38.70


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

refractive index

n20/D 1.531 (lit.)

bp

99-101 °C/12 mmHg (lit.)

mp

21 °C (lit.)

density

1.227 g/mL at 25 °C (lit.)

functional group

chloro
ester

SMILES string

COC(=O)c1cccc(Cl)c1

InChI

1S/C8H7ClO2/c1-11-8(10)6-3-2-4-7(9)5-6/h2-5H,1H3

InChI key

XRDRKVPNHIWTBX-UHFFFAOYSA-N

General description

The (1+1) Resonance Enhanced Multiphoton Ionisation (REMPI) spectrum and the fluorescence excitation spectrum of methyl 3-chlorobenzoate has been measured in the supersonic free jet[1]. Reaction of methyl 3-chlorobenzoate with lithium diisobutyl-t-butoxyaluminum hydride (LDBBA) has been investigated[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laser jet spectra of methyl 3-chlorobenzoate.
Hirano A, et al.
Journal of Molecular Structure, 701(1), 9-11 (2004)
Lithium diisobutylpiperidinohydroaluminate (LDBPA) as a mild and efficient reducing agent for the conversion of ester to aldehyde.
Ahn JH, et al.
Bull. Korean Chem. Soc., 26, 377-378 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service