Skip to Content
Merck
All Photos(1)

Key Documents

316598

Sigma-Aldrich

Isobutyl (R)-(+)-lactate

97%

Synonym(s):

(+)-Isobutyl D-lactate, R-(+)-Lactic acid isobutyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€359.00

€359.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
€359.00

About This Item

Linear Formula:
CH3CH(OH)CO2CH2CH(CH3)2
CAS Number:
Molecular Weight:
146.18
Beilstein:
1721591
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

€359.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

liquid

optical activity

[α]19/D +15°, neat

refractive index

n20/D 1.419 (lit.)

bp

73 °C/13 mmHg (lit.)

density

0.971 g/mL at 25 °C (lit.)

functional group

ester
hydroxyl

SMILES string

CC(C)COC(=O)[C@@H](C)O

InChI

1S/C7H14O3/c1-5(2)4-10-7(9)6(3)8/h5-6,8H,4H2,1-3H3/t6-/m1/s1

InChI key

WBPAQKQBUKYCJS-ZCFIWIBFSA-N

Other Notes

remainder mainly isobutanol

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Repellent effect of Mannich bases, derivatives of lactic acid esters].
L G Kovalenko et al.
Meditsinskaia parazitologiia i parazitarnye bolezni, (1)(1), 44-48 (1987-01-01)
J J Clary et al.
Regulatory toxicology and pharmacology : RTP, 27(2), 88-97 (1998-07-22)
Lactate esters have an oral LD50 greater than 2000 mg/kg and the inhalation LC50 is generally above 5000 mg/m3 and they may be potential eye and skin irritants, but not skin sensitizers. No evidence of teratogenicity or maternal toxicity was
Direct separation of alpha-hydroxycarboxylic acid ester enantiomers by gas chromatography with optically active copper(II) complexes.
N Oi et al.
Journal of chromatography, 202(2), 305-308 (1980-12-19)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service