Skip to Content
Merck
All Photos(1)

Documents

8.22100

Sigma-Aldrich

(S)-(-)-Ethyl lactate

for synthesis

Synonym(s):

(S)-(-)-Ethyl lactate, L(-)-Lactic acid ethyl ester, (S)-(-)-2-Hydroxypropionic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)COOC2H5
CAS Number:
Molecular Weight:
118.13
MDL number:
UNSPSC Code:
12352108
EC Index Number:
211-694-1
NACRES:
NA.22

vapor pressure

1.6 hPa ( 20 °C)

Quality Level

Assay

≥99% (GC)

form

liquid

autoignition temp.

400 °C

potency

>2000 mg/kg LD50, oral (Rat)

expl. lim.

1.5-16.4 % (v/v)

pH

4 (20 °C, 50 g/L in H2O)

kinematic viscosity

2.7 cSt(25 °C)

bp

154 °C/1013 hPa

mp

-25 °C

transition temp

flash point 53 °C

density

1.03 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C5H10O3/c1-3-8-5(7)4(2)6/h4,6H,3H2,1-2H3/t4-/m0/s1

InChI key

LZCLXQDLBQLTDK-BYPYZUCNSA-N

Related Categories

Application



  • Evaluation of operational parameters for ethyl lactate production using reactive distillation process: This study explores the environmentally benign production of ethyl lactate, highlighting its use as a bio-based solvent for chemical transformations (A Komesu et al., 2015).


  • Influence of sequential fermentation with Torulaspora delbrueckii and Saccharomyces cerevisiae on wine quality: Discusses the production of ethyl lactate and its effect on wine quality, providing insights into fermentation processes and flavor enhancement (I Loira et al., 2014).


  • Titania-catalysed oxidative dehydrogenation of ethyl lactate: Examines a catalytic process for converting ethyl lactate to ethyl pyruvate, emphasizing green chemistry applications (EV Ramos-Fernandez et al., 2014).


  • Green Sustainable Process for Chemical and Environmental Engineering and Science: Focuses on the role of ethyl lactate in organic synthesis, showcasing its effectiveness in various chemical reactions (AV Dolzhenko, 2021).


  • In Situ Analysis of Metabolic Characteristics Reveals the Key Yeast in the Spontaneous and Solid-State Fermentation Process of Chinese Light-Style Liquor: Investigates the influence of ethyl lactate on fermentation, particularly its impact on flavor profiles in beverage production (Y Kong et al., 2014).


Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.032 - 1.034
Spec. rotation [α²0/D (undiluted): -11.0 - -9.5 °
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

128.8 °F - closed cup

Flash Point(C)

53.8 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service