Skip to Content
Merck
All Photos(3)

Key Documents

310476

Sigma-Aldrich

(R)-(−)-5-Oxo-2-tetrahydrofurancarboxylic acid

98%

Synonym(s):

(R)-γ-Carboxy-γ-butyrolactone, (R)-5-Oxotetrahydro-2-furancarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€87.60
5 G
€252.00

€87.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€87.60
5 G
€252.00

About This Item

Empirical Formula (Hill Notation):
C5H6O4
CAS Number:
Molecular Weight:
130.10
Beilstein:
1424499
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€87.60


Please contact Customer Service for Availability

Request a Bulk Order

Assay

98%

form

solid

optical activity

[α]20/D −14°, c = 5 in methanol

optical purity

ee: 97% (GLC)

bp

165-167 °C/0.3 mmHg (lit.)

mp

71-73 °C (lit.)

functional group

carboxylic acid
ester

SMILES string

OC(=O)[C@H]1CCC(=O)O1

InChI

1S/C5H6O4/c6-4-2-1-3(9-4)5(7)8/h3H,1-2H2,(H,7,8)/t3-/m1/s1

InChI key

QVADRSWDTZDDGR-GSVOUGTGSA-N

General description

(R)-(−)-5-Oxo-2-tetrahydrofurancarboxylic acid belongs to the class of furans, considered as heterocyclic building blocks. It is used as an HPLC derivatization reagent for UV/Vis detection.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shinji K Strain et al.
Rapid communications in mass spectrometry : RCM, 33(17), 1401-1409 (2019-05-31)
2-Hydroxyglutarate (2-hg) exists as enantiomers and can readily undergo cyclization to its lactone. Gas chromatography/electron ionization mass spectrometry (GC/EI-MS) has been used to separate 2-hg enantiomers in bodily fluids but the assay cannot simultaneously measure cyclic and acylic 2-hg enantiomers.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service