Skip to Content
Merck
All Photos(3)

Key Documents

P65800

Sigma-Aldrich

3-Pyridylacetic acid hydrochloride

98%

Synonym(s):

3-Pyridineacetic acid hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€26.30
5 G
€38.30
25 G
€86.50

€26.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€26.30
5 G
€38.30
25 G
€86.50

About This Item

Empirical Formula (Hill Notation):
C7H7NO2 · HCl
CAS Number:
Molecular Weight:
173.60
Beilstein:
3696630
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€26.30


Please contact Customer Service for Availability

Request a Bulk Order

Assay

98%

form

crystals

mp

161-163 °C (lit.)

SMILES string

Cl[H].OC(=O)Cc1cccnc1

InChI

1S/C7H7NO2.ClH/c9-7(10)4-6-2-1-3-8-5-6;/h1-3,5H,4H2,(H,9,10);1H

InChI key

XVCCOEWNFXXUEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zahra Salmasi et al.
Biotechnology progress, 36(6), e3025-e3025 (2020-05-16)
Cell-based delivery system is a promising strategy to protect therapeutic agents from the immune system and provide targeted delivery. Mesenchymal stem cells (MSCs) have recently been introduced as an encouraging vehicle in cell-based gene therapy due to their unique features
Maryam Hashemi et al.
Materials science & engineering. C, Materials for biological applications, 61, 791-800 (2016-02-04)
Poly-(amidoamine) (PAMAM) and poly-(propylenimine) (PPI) are the two most widely investigated dendrimers for drug and gene delivery. In order to enhance DNA transfection activity of these dendrimers, generation 3 and 4 PAMAM and generation 4 and 5 PPI were modified
Zahra Salmasi et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 96, 76-88 (2015-07-26)
Branched polyethylenimine (PEI) is extensively used as a polycationic non-viral vector for gene delivery. Polyplexes formed with PEI are believed to be released from endocytotic vesicles by the osmotic burst mechanism in the rate-limiting step in transfection. Increasing the buffering

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service