Skip to Content
Merck
All Photos(1)

Key Documents

307572

Sigma-Aldrich

Propionyl bromide

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€54.60
25 G
€190.00

€54.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€54.60
25 G
€190.00

About This Item

Linear Formula:
CH3CH2COBr
CAS Number:
Molecular Weight:
136.98
Beilstein:
1736651
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€54.60


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.455 (lit.)

bp

103-104 °C (lit.)

density

1.521 g/mL at 25 °C (lit.)

functional group

acyl bromide

SMILES string

CCC(Br)=O

InChI

1S/C3H5BrO/c1-2-3(4)5/h2H2,1H3

InChI key

RIBFXMJCUYXJDZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular structure, composition, and energy and entropy differences between conformers of propionyl bromide as determined by gas-phase electron diffraction.
The Journal of Physical Chemistry, 91(15), 3977-3981 (1987)
Aniruddha Pal et al.
International journal of biological macromolecules, 119, 954-961 (2018-08-05)
Dextrin and poly (N-vinyl caprolactam) based amphiphilic graft copolymer has recently been developed using RAFT polymerization. The prepared graft copolymer has been characterized in details using FTIR and 1H NMR spectral analyses, GPC, TGA, FESEM, TEM and DLS analyses. GPC
Hasmik Grigoryan et al.
Carcinogenesis, 39(5), 661-668 (2018-03-15)
Although benzene has long been recognized as a cause of human leukemia, the mechanism by which this simple molecule causes cancer has been problematic. A complicating factor is benzene metabolism, which produces many reactive intermediates, some specific to benzene and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service