Skip to Content
Merck
All Photos(1)

Key Documents

8.07706

Sigma-Aldrich

Silicon tetrachloride

for synthesis

Synonym(s):

Silicon tetrachloride, Silicon(IV) chloride, Tetrachlorosilane

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€61.00
1 L
€199.00

€61.00


Estimated to ship on24 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
€61.00
1 L
€199.00

About This Item

Empirical Formula (Hill Notation):
Cl4Si
CAS Number:
Molecular Weight:
169.90
MDL number:
UNSPSC Code:
12352307
EC Index Number:
233-054-0
NACRES:
NA.22

€61.00


Estimated to ship on24 April 2025Details


Request a Bulk Order

vapor pressure

257 hPa ( 20 °C)

Quality Level

form

liquid

pH

1 (5 g/L in H2O)

bp

57 °C/1013 hPa

mp

-70 °C

density

1.48 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Si](Cl)(Cl)(Cl)Cl

InChI

1S/Cl4Si/c1-5(2,3)4

InChI key

FDNAPBUWERUEDA-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 99.0 %

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service