Skip to Content
Merck
All Photos(1)

Documents

301264

Sigma-Aldrich

Ethylamine

97%

Synonym(s):

Aminoethane, Monoethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5NH2
CAS Number:
Molecular Weight:
45.08
Beilstein:
505933
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

vapor density

1.56 (15 °C, vs air)

vapor pressure

874 mmHg ( 20 °C)

Assay

97%

autoignition temp.

721 °F

expl. lim.

14 %
3.5-14 %

bp

16.6 °C (lit.)

mp

-81 °C (lit.)

density

0.680 g/mL at 20 °C (lit.)
0.689 g/mL at 25 °C (lit.)

SMILES string

CCN

InChI

1S/C2H7N/c1-2-3/h2-3H2,1H3

InChI key

QUSNBJAOOMFDIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylamine, also known as ethanamine is commonly employed as a precursor in the synthesis of organic compounds. It is a corrosive primary amine, which is found naturally in many vegetables and fruits. It is also present as a constituent of tobacco smoke. Ethylamine is known to affect the lungs and kidney when a person is exposed to it.

Application

Ethylamine can be used as a reagent:
  • In the hydroamination reaction to prepare ethylamine derivatives by addition reaction with alkynes using the Ind2TiMe2 catalyst.
  • To prepare Schiff base ligands, which are reacted with various metal cations to obtain mono- or binuclear ethylamine metal complexes.
  • To synthesize ethylamine-modified montmorillonite, which is used as a novel material for the removal of radiocesium from radioactive wastewater.

Packaging

Supplied in a Sure/Pac cylinder and has a 316 stainless steel needle valve with a male 1/4" NPTF outlet thread installed . Before using the cylinder, ensure that the valve is closed, then remove the galvanized steel hex cap that seals the outlet valve.

Compatible with the following:
  • Aldrich® Sure/Pac station for liquefied gases Z566446
  • PTFE Sealing tape Z104388 or Z221880
  • Stainless steel hose adapter Z146838
  • Aldrich® mini gas regulator Z513547

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Gas 1 - Press. Gas Liquefied gas - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

-34.6 °F - closed cup

Flash Point(C)

-37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ethylamine
Encyclopedia of Toxicology (2014)
Amines, aliphatic
K Eller, et al.
Ullmann's Encyclopedia of Industrial Chemistry (2000)
Synthesis and Spectroscopic Studies of a Novel Schiff Base Derived from o-Acetoacetylphenol and Ethylamine and Its Metal Complexes
Salib KAR, et al.
Synth. React. Inorg. Met.-Org. Chem. , 33(9), 1667-1687 (2003)
Ind2TiMe2-Catalyzed Addition of Methyl-and Ethylamine to Alkynes
Marcsekov'a K, et al.
European Journal of Organic Chemistry, 2005(22), 4843-4851 (2005)
Evaluation of Cs+ removal from aqueous solution by adsorption on ethylamine-modified montmorillonite
Long H, et al.
Chemical Engineering Journal, 225(9), 237-244 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service