Skip to Content
Merck
All Photos(2)

Key Documents

8.20024

Sigma-Aldrich

Ethylamine

(70% aqueous solution) for synthesis

Synonym(s):

Aminoethane, Monoethylamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€41.90
2.5 L
€64.90

€41.90


Estimated to ship on24 March 2025Details


Request a Bulk Order

Select a Size

Change View
1 L
€41.90
2.5 L
€64.90

About This Item

UNSPSC Code:
12352116
NACRES:
NA.22

€41.90


Estimated to ship on24 March 2025Details


Request a Bulk Order

vapor pressure

460 hPa ( 20 °C)

Quality Level

200
300

form

liquid

autoignition temp.

375 °C

potency

400 mg/kg LD50, oral (Rat)
265 mg/kg LD50, skin (Rabbit)

expl. lim.

3.0-12.8 % (v/v)

pH

>14 (20 °C in H2O)

bp

39 °C/1013 hPa

mp

<-70 °C

transition temp

flash point -26 °C

density

0.81 g/cm3 at 20 °C

storage temp.

2-30°C

Application

  • Exploring the Biomedical Potential of Terpenoid Alkaloids: Discusses the synthesis of terpenoid alkaloids, including the introduction of ethylamine, highlighting their structures and biomedical applications (X Wang, J Xin, L Sun, Y Sun, Y Xu, F Zhao, C Niu, S Liu - Molecules, 2024). Link.

Analysis Note

Assay (acidimetric): 68.0 - 72.0 %
Density (d 20 °C/ 4 °C): 0.803 - 0.812

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(C)

-26 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service