Skip to Content
Merck
All Photos(1)

Key Documents

300373

Sigma-Aldrich

2,3-Dibromopropionyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CHBrCOCl
CAS Number:
Molecular Weight:
250.32
Beilstein:
1749801
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.542 (lit.)

bp

191-193 °C (lit.)

density

2.181 g/mL at 25 °C (lit.)

SMILES string

ClC(=O)C(Br)CBr

InChI

1S/C3H3Br2ClO/c4-1-2(5)3(6)7/h2H,1H2

InChI key

HWKWYDXHMQQDQJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,3-Dibromopropionyl chloride was used in synthesis of:
  • series of 2,3-diaminopropionanilides
  • enantiomers of 1,4-dideoxy-1,4-iminolyxitol and 1,4-dideoxy-1,4-iminoribitol
  • acromelic acid A

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiral synthesis of both enantiomers of 1, 4-dideoxy-1, 4-immolyxitol and 1, 4-dideoxy-1, 4-iminoribitol.
Takano S, et al.
Tetrahedron Asymmetry, 3(6), 681-684 (1992)
Concise enantioselective synthesis of acromelic acid A.
Takano S, et al.
Journal of the American Chemical Society, 109(18), 5523-5524 (1987)
P A Tenthorey et al.
Journal of medicinal chemistry, 24(9), 1059-1063 (1981-09-01)
A series of 2,3-diaminopropionanilides was synthesized by acylation of mono- and disubstituted aniline derivatives with 2,3-dibromopropionyl chloride and subsequent amination with the appropriate secondary amines. The target compounds were evaluated in mice for antiarrhythmic efficacy against chloroform-induced tachycardia and for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service