Skip to Content
Merck
All Photos(1)

Key Documents

167118

Sigma-Aldrich

Ethyl 4-bromobutyrate

95%

Synonym(s):

γ-Bromobutyric acid ethyl ester, 1-Bromo-3-(carboethoxy)propane, 3-(Ethoxycarbonyl)propyl bromide, 4-Bromobutanoic acid ethyl ester, 4-Bromobutyric acid ethyl ester, Ethyl γ-bromobutyrate, Ethyl 4-bromobutanoate

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€46.60
250 G
€197.00

€46.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€46.60
250 G
€197.00

About This Item

Linear Formula:
Br(CH2)3COOC2H5
CAS Number:
Molecular Weight:
195.05
Beilstein:
1749700
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€46.60


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.456 (lit.)

bp

80-82 °C/10 mmHg (lit.)

density

1.363 g/mL at 25 °C (lit.)

functional group

bromo
ester

SMILES string

CCOC(=O)CCCBr

InChI

1S/C6H11BrO2/c1-2-9-6(8)4-3-5-7/h2-5H2,1H3

InChI key

XBPOBCXHALHJFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl 4-bromobutyrate was used in Enzyme-linked immunosorbent assay for zilpaterol[1]. It was used in the synthesis of isosteric thieno[2,3-b]-azepin-4-ones, having anti-tumor activity[2]. It was also used in the synthesis of poly(2,6-dimethyl-1,4-phenylene oxide)[3].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R F Koebel et al.
Journal of medicinal chemistry, 18(2), 192-194 (1975-02-11)
In view of the antitumor activity reported for 7,8-dimethylbenzo[b]azepine-2,5-dione, new isosteric thieno[2,3-b]-azepin-4-ones have been prepared by a Dieckmann ring closure reaction. Substituted 2-amino-3-carbethoxythiophenes were tosylated, or benzoylated, and the corresponding sodium salt was alkylated with ethyl 4-bromobutyrate. The resulting product
Weilin L Shelver et al.
Journal of agricultural and food chemistry, 52(8), 2159-2166 (2004-04-15)
Zilpaterol is an beta-adrenergic agonist approved for use in cattle in South Africa and Mexico as a growth promoter. It is not currently approved for use in the EU, USA, or Asia. Here, we report the development of an ELISA
Xingdong Yang et al.
Scientific reports, 8(1), 16680-16680 (2018-11-14)
A lateral flow immunochromatographic strip test (LFIST) based on a competitive format was developed for rapid and sensitive on-site detection of oseltamivir phosphate (OP) residues in poultry product. The sensitivity (half inhibitory concentration, IC50) of the LFIST in the detection
Prithidipa Sahoo et al.
Bioorganic chemistry, 71, 315-324 (2017-03-14)
Naphthyridine-based fluorescent probe H1 was synthesized and characterized for the quantification and selective detection of Uric Acid (UA) in live cell. In presence of UA, H1 forms the specific host-guest complex mainly through intermolecular hydrogen bonding and aromatic stacking which
Kai Yao et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1080, 50-58 (2018-02-25)
An immunoaffinity clean-up material based on a monoclonal antibody (mAb) has been prepared for concentrating and purifying bisphenol A (BPA) in 14 kinds of foodstuffs at trace level. Haptens and immunogen of bisphenol A have been synthesized and comprehensively characterized.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service