Skip to Content
Merck
All Photos(1)

Key Documents

298719

Sigma-Aldrich

Methyl 2-oxo-1-cycloheptanecarboxylate

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€81.20
25 G
€385.00

€81.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€81.20
25 G
€385.00

About This Item

Linear Formula:
C7H11(O)CO2CH3
CAS Number:
Molecular Weight:
170.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€81.20


Please contact Customer Service for Availability

Request a Bulk Order

Assay

99%

refractive index

n20/D 1.474 (lit.)

bp

112-114 °C/10 mmHg (lit.)

density

1.09 g/mL at 25 °C (lit.)

functional group

ester
ketone

SMILES string

COC(=O)C1CCCCCC1=O

InChI

1S/C9H14O3/c1-12-9(11)7-5-3-2-4-6-8(7)10/h7H,2-6H2,1H3

InChI key

BLNHITXSJKCQGZ-UHFFFAOYSA-N

Application

Methyl 2-oxo-1-cycloheptanecarboxylate was used in preparation of:
  • hydroxycoumarins via Pechmann condensation with polyphenols[1]
  • 3-hydroxy-6,7,8,9,10,11-hexahydrocyclohepta[c]chromen-6-one[1]
  • 3-hydroxy-4-methyl-6,7,8,9,10,11-hexahydrocyclohepta[c]chromen-6-one[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modified coumarins. 12. Synthesis of 3, 4-cycloannelated coumarin ?-d-glucopyranosides.
Garazd YL, et al.
Chemistry of Natural Compounds, 40(1), 6-12 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service