Skip to Content
Merck
All Photos(2)

Key Documents

168106

Sigma-Aldrich

Methyl 2-oxocyclopentanecarboxylate

95%

Synonym(s):

2-Methoxycarbonylcyclopentanone, Methyl cyclopentanone-2-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€27.10
100 G
€31.40

€27.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€27.10
100 G
€31.40

About This Item

Linear Formula:
(O=)C5H7CO2CH3
CAS Number:
Molecular Weight:
142.15
Beilstein:
971717
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€27.10


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

95%

refractive index

n20/D 1.456 (lit.)

bp

105 °C/19 mmHg (lit.)

density

1.145 g/mL at 25 °C (lit.)

functional group

ester
ketone

storage temp.

2-8°C

SMILES string

COC(=O)C1CCCC1=O

InChI

1S/C7H10O3/c1-10-7(9)5-3-2-4-6(5)8/h5H,2-4H2,1H3

InChI key

PZBBESSUKAHBHD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kinetics of complexation of methyl 2-oxocyclopentanecarboxylate with Cr(III) in aqueous solution has been investigated[1].

Application

Methyl 2-oxocyclopentanecarboxylate was used in the synthesis of:
  • 1,2-disubstituted imidazolylmethylcyclopentanol derivatives[2]
  • methyl 1-(4-bromobutyl)-2-oxocyclopentanecarboxylate[3]
  • methyl 1-(4-iodobutyl)-2-oxocyclopentanecarboxylate[3]
  • ethyl 1-(3-bromopropyl)-2-oxocyclohexanecarboxylate[3]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nickel (I) Salen-Catalyzed Reduction of 1-Haloalkyl-2-oxocycloalkanecarboxylates Three-and Four-Carbon Ring Expansions.
Mubarak MS, et al.
Journal of the Electrochemical Society, 154(11), F205-F210 (2007)
Kinetics of monochelation of aqueous chromium (III) by methyl 2-oxocyclopentanecarboxylate.
Rojas A, et al.
Journal of Physical Organic Chemistry, 13(2), 97-104 (2000)
A Kato et al.
Chemical & pharmaceutical bulletin, 43(12), 2152-2158 (1995-12-01)
Two series of 1,2-disubstituted imidazolylmethylcyclopentanol derivatives (5a-d, 10a-d) were prepared by using easily available methyl 2-oxocyclopentanecarboxylate as the starting material. Evaluation of the aromatase inhibitory activities in vitro was performed. Their activities were compared with those of a steroidal aromatase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service