Skip to Content
Merck
All Photos(1)

Documents

297763

Sigma-Aldrich

4-Pyridazinecarboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N2O2
CAS Number:
Molecular Weight:
124.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

244.2 °C (dec.) (lit.)

SMILES string

OC(=O)c1ccnnc1

InChI

1S/C5H4N2O2/c8-5(9)4-1-2-6-7-3-4/h1-3H,(H,8,9)

InChI key

JUSIWJONLKBPDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Surface electrochemical studies of 4-pyridazinecarboxylic acid adsorbed at Pt(111) surfaces by electron energy-loss and Auger spectroscopy has been reported.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multinitrogen heteroaromatics studied at Pt (111) surfaces by EELS, Auger spectroscopy, and electrochemistry: pyrazine, pyrimidine, pyridazine, 1, 3, 5-triazine, and their carboxylic acid derivatives.
Chaffins SA, et al.
Langmuir, 6(7), 1273-1281 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service