Skip to Content
Merck
All Photos(2)

Key Documents

282391

Sigma-Aldrich

Methylsulfamic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NHSO3H
CAS Number:
Molecular Weight:
111.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

191 °C (dec.) (lit.)

functional group

amine

SMILES string

CNS(O)(=O)=O

InChI

1S/CH5NO3S/c1-2-6(3,4)5/h2H,1H3,(H,3,4,5)

InChI key

MYMDOKBFMTVEGE-UHFFFAOYSA-N

Application

Methylsulfamic acid has been used in the preparation of nitroaroyl- or nitroarylsulphonyl-sulphamic acids via condensation reaction with certain nitroaroyl and nitroarylsulphonyl chlorides[1].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aroyl-and arylsulphonyl-sulphamic acids.
<BIG>Beech WF. </BIG>
J. Chem. Soc. Sect. C, 3, 515-516 (1970)
Akifumi Kogame et al.
Xenobiotica; the fate of foreign compounds in biological systems, 47(3), 255-266 (2016-05-27)
1. Following oral administration of [

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service