Skip to Content
Merck
All Photos(1)

Key Documents

278246

Sigma-Aldrich

3-Cyano-6-methyl-2(1H)-pyridinone

97%

Synonym(s):

1,2-Dihydro-6-methyl-2-oxo-3-pyridinecarbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N2O
CAS Number:
Molecular Weight:
134.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

293-295 °C (lit.)

functional group

nitrile

SMILES string

CC1=CC=C(C#N)C(=O)N1

InChI

1S/C7H6N2O/c1-5-2-3-6(4-8)7(10)9-5/h2-3H,1H3,(H,9,10)

InChI key

FIMGYEKEYXUTGD-UHFFFAOYSA-N

General description

Supramolecular structures of coordination copper(I) compounds of 3-cyano-6-methyl-2(1H)-pyridinone has been investigated[1].

Application

3-Cyano-6-methyl-2(1H)-pyridinone has been used in the preparation of the N3′-pyridyl thiamine, a potent in vitro thiamine antagonist[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Construction of three-dimensional supramolecular coordination copper (I) compounds with channel structures hosting a variety of anions by changing the hydrogen-bonding mode and distances.
Munakata M, et al.
Journal of the American Chemical Society, 118(13), 3117-3124 (1996)
Allen A Thomas et al.
Bioorganic & medicinal chemistry letters, 18(6), 2206-2210 (2008-02-13)
Tumor cells extensively utilize the pentose phosphate pathway for the synthesis of ribose. Transketolase is a key enzyme in this pathway and has been suggested as a target for inhibition in the treatment of cancer. In a pharmacodynamic study, nude

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service