Skip to Content
Merck
All Photos(1)

Documents

275700

Sigma-Aldrich

2,3-Dichlorohexafluoro-2-butene, mixture of cis and trans

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C(Cl)=C(Cl)CF3
CAS Number:
Molecular Weight:
232.94
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

8 (vs air)

vapor pressure

4.42 psi ( 20 °C)

Assay

99%

refractive index

n20/D 1.3458 (lit.)

bp

66-68 °C (lit.)

density

1.605 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C(\Cl)=C(/Cl)C(F)(F)F

InChI

1S/C4Cl2F6/c5-1(3(7,8)9)2(6)4(10,11)12/b2-1+

InChI key

XDIDQEGAKCWQQP-OWOJBTEDSA-N

General description

The pure cis- and trans-isomers of 2,3-dichlorohexafluoro-2-butene were separated when 2,3-dichlorohexafluoro-2-butene (mixture of cis and trans) reacted with dry diglyme and sodium borohydride and was hydolyzed. The isomers were purified by preparative vapor phase chromatography technique.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

THE PREPARATION AND CHARACTERIZATION OF cis-AND trans-2-HYDRO-3-CHLOROHEXAFLUORO-2-BUTENE.
Burton DJ, et al.
Canadian Journal of Chemistry, 44(5), 635-636 (1966)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service