Skip to Content
Merck
All Photos(1)

Key Documents

272507

Sigma-Aldrich

2-Chloro-2′,4′-difluoroacetophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3COCH2Cl
CAS Number:
Molecular Weight:
190.57
Beilstein:
1943217
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 272507, please contact your local Merck office or dealer. Contact Technical Service

Assay

98%

form

solid

mp

44-48 °C (lit.)

functional group

chloro
fluoro
ketone

SMILES string

Fc1ccc(c(F)c1)C(=O)CCl

InChI

1S/C8H5ClF2O/c9-4-8(12)6-2-1-5(10)3-7(6)11/h1-3H,4H2

InChI key

UENGBOCGGKLVJJ-UHFFFAOYSA-N

Gene Information

human ... PTPN6(5777)

Looking for similar products? Visit Product Comparison Guide

Application

2-Chloro-2′,4′-difluoroacetophenone was used in the synthesis of allyl alcohol[1].

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective synthesis of the optical isomers of broad-spectrum orally active antifungal azoles, SCH 42538 and SCH 45012.
Bioorganic & Medicinal Chemistry Letters, 4(16), 2023-2028 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service