
2-Chloro-2′,4′-difluoroacetophenone

Linear Formula:
F2C6H3COCH2Cl
CAS Number:
Molecular Weight:
190.57
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing