267988
Isodrine
Synonym(s):
1,2,3,4,10,10-Hexachloro-1,4,4a,5,8,8a-hexahydro-endo,endo-1,4:5,8-dimethanonaphthalene
Select a Size
Select a Size
About This Item
Recommended Products
form
solid
mp
239-241 °C (lit.)
functional group
chloro
SMILES string
[H][C@@]12C3CC(C=C3)[C@]1([H])[C@]4(Cl)C(Cl)=C(Cl)[C@@]2(Cl)C4(Cl)Cl
InChI
1S/C12H8Cl6/c13-8-9(14)11(16)7-5-2-1-4(3-5)6(7)10(8,15)12(11,17)18/h1-2,4-7H,3H2/t4-,5+,6-,7+,10-,11+
InChI key
QBYJBZPUGVGKQQ-DIFDVCDBSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service