Skip to Content
Merck
All Photos(1)

Key Documents

31041

Supelco

4,4′-DDT

PESTANAL®, analytical standard

Synonym(s):

1,1,1-Trichloro-2,2-bis(4-chlorophenyl)ethane, 1,1-Bis(4-chlorophenyl)-2,2,2-trichloroethane

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€86.20

€86.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€86.20

About This Item

Linear Formula:
(ClC6H4)2CHCCl3
CAS Number:
Molecular Weight:
354.49
Beilstein:
1882657
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€86.20


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

107-110 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Clc1ccc(cc1)C(c2ccc(Cl)cc2)C(Cl)(Cl)Cl

InChI

1S/C14H9Cl5/c15-11-5-1-9(2-6-11)13(14(17,18)19)10-3-7-12(16)8-4-10/h1-8,13H

InChI key

YVGGHNCTFXOJCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,4′-DDT (Dichlorodiphenyltrichloroethane) is a notorious organochlorine pesticide. It contains 77% p′p′-DDT, 15% o′p′-DDT, 4% p′p′-DDE (Dichlorodiphenyltrichloroethylene) and less than 1% o′p′-DDD (Dichlorodiphenyldichloroethane), p′p′-DDD and o′p′-DDE.

Application

4,4′-DDT may be used as a reference standard for the determination of the chlorinated pesticide:
  • In water by solid-phase microextraction followed by gas chromatography (GC) coupled with an electron capture detector.[1]
  • In human biological samples by gas chromatography (GC) with mass spectrometry (MS) detector in tandem mode.[2]
It may also be used as a standard to spike milk samples during extraction method, further followed by column chromatography to determine organochlorine pollutants.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT RE 1 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Determination of chlorinated pesticides in water by SPME/GC.
Boussahel R, et al.
Water Research, 36(7), 1909- 1911 (2002)
Determination of organochlorine compounds in human biological samples by GC-MS/MS.
Frias MM, et al.
Biomedical Chromatography, 18(2), 102- 111 (2004)
Xiao-Ting Jin et al.
Toxicology letters, 225(1), 158-166 (2013-12-21)
Dichlorodiphenyltrichloroethane (DDT) is a persistent organic pollutant, involved in the progression of many cancers, including liver cancer. However, the underlying mechanism(s) of DDT, especially how low doses DDT cause liver cancer, is poorly understood. In this study, we evaluated the
Florian Thevenon et al.
Chemosphere, 90(9), 2444-2452 (2012-12-04)
The (137)Cs and (210)Pb dating of a 61-cm long sediment core retrieved from a drinking water reservoir (Lake Brêt) located in Switzerland revealed a linear and relatively high sedimentation rate (~1 cm year(-1)) over the last decades. The continuous centimeter
Katarzyna Kulma et al.
PloS one, 8(3), e58322-e58322 (2013-03-14)
Insecticide-resistance threatens the control of mosquito-borne diseases like malaria or dengue fever. To ensure sustainable vector control we need a full understanding of the factors driving the evolution of resistance. We test the hypothesis that the expression of insecticide-resistance depends

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service