Skip to Content
Merck
All Photos(1)

Key Documents

265136

Sigma-Aldrich

2-Bromovaleric acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(Br)CO2H
CAS Number:
Molecular Weight:
181.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.4709 (lit.)

bp

132-136 °C/25 mmHg (lit.)

density

1.381 g/mL at 25 °C (lit.)

functional group

bromo
carboxylic acid

SMILES string

CCCC(Br)C(O)=O

InChI

1S/C5H9BrO2/c1-2-3-4(6)5(7)8/h4H,2-3H2,1H3,(H,7,8)

InChI key

WMFATTFQNRPXBQ-UHFFFAOYSA-N

Application

2-Bromovaleric acid was used in the solid-phase synthesis of 2,4,5-trisubstituted thiomorpholine-3-ones.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Nefzi et al.
Bioorganic & medicinal chemistry letters, 8(17), 2273-2278 (1999-01-05)
Modified dipeptides have been used successfully for the generation of a variety of small organic and heterocyclic combinatorial libraries, including linear urea, polyamine, hydantoin, thiohydantoin, cyclic urea, cyclic thiourea and bicyclic guanidine. The synthesis and screening results for a number
Diego Méndez et al.
Biochemical pharmacology, 183, 114341-114341 (2020-11-17)
Platelets are the smallest blood cells, and their activation (platelet cohesion or aggregation) at sites of vascular injury is essential for thrombus formation. Since the use of antiplatelet therapy is an unsolved problem, there are now focused and innovative efforts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service