Skip to Content
Merck
All Photos(2)

Documents

306851

Sigma-Aldrich

2-Bromo-2-methylpropionic acid

98%

Synonym(s):

α-Bromoisobutyric acid, 2-Bromo-2-methylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CBrCOOH
CAS Number:
Molecular Weight:
167.00
Beilstein:
1745543
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

bp

198-200 °C (lit.)

mp

44-47 °C (lit.)

SMILES string

CC(C)(Br)C(O)=O

InChI

1S/C4H7BrO2/c1-4(2,5)3(6)7/h1-2H3,(H,6,7)

InChI key

XXSPGBOGLXKMDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The multifunctional silica colloids were prepared by coating them with 2-bromo-2-methylpropionic acid stabilized quantum dots.

Application

2-Bromo-2-methylpropionic acid was used in the synthesis of dextran macroinitiator for atom transfer radical polymerization (ATRP) by partial esterification of hydroxyl group of the polysaccharide.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fan Li et al.
Carbohydrate polymers, 99, 728-735 (2013-11-28)
A new class of biodegradable, biomimetic and thermo-responsive dextran/synthetic glycopolymer biohybrids (dextran-graft-poly(lactobionamidoethyl methacrylate)-block-poly(di(ethylene glycol) methyl ether methacrylate), dextran-g-(PLAMA-b-PDEGMA)), was synthesized by the direct atom transfer radical polymerization (ATRP) of unprotected lactobionamidoethyl methacrylate (LAMA) glycomonomer and di(ethylene glycol) methyl ether methacrylate
Miseon Yoon et al.
ACS nano, 5(7), 5417-5426 (2011-06-22)
We demonstrate the successful preparation of multifunctional silica colloids by coating with 2-bromo-2-methylpropionic acid (BMPA)-stabilized quantum dots (BMPA-QDs) and BMPA-stabilized iron oxide particles (BMPA-Fe(3)O(4)), along with amine-functionalized poly(amidoamine) (PAMA) dendrimers, using layer-by-layer (LbL) assembly based on a nucleophilic substitution (NS)
Raj Kumar Roy et al.
Nature communications, 6, 7237-7237 (2015-05-27)
Biopolymers such as DNA store information in their chains using controlled sequences of monomers. Here we describe a non-natural information-containing macromolecule that can store and retrieve digital information. Monodisperse sequence-encoded poly(alkoxyamine amide)s were synthesized using an iterative strategy employing two
Peng Yan et al.
Acta biomaterialia, 10(8), 3786-3794 (2014-05-13)
The fabrication of highly efficient nonviral gene carriers with low cytotoxicity remains a challenge in gene therapy. This paper reports a facile strategy to combine the advantages of gold nanorods (Au NRs) and polycations through surface functionalization. Different Au NR
Fabio Salvatore Palumbo et al.
Macromolecular bioscience, 15(11), 1605-1615 (2015-07-03)
Two derivatives of hyaluronic acid (HA) have been synthesized by atom transfer radical polymerization (ATRP), starting from an ethylenediamino HA derivative (HA-EDA) and by using diethylaminoethyl methacrylate (DEAEMA) as a monomer for polymerization. Both samples, indicated as HA-EDA-pDEAEMA a and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service