253634
5-DOXYL-stearic acid, free radical
Synonym(s):
2-(3-Carboxypropyl)-4,4-dimethyl-2-tridecyl-3-oxazolidinyloxy, free radical, 5-Doxylstearate, 5-SASL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
solid
mp
51-53 °C (lit.)
storage temp.
2-8°C
SMILES string
CCCCCCCCCCCCCC1(CCCC(O)=O)OCC(C)(C)N1[O]
InChI
1S/C22H42NO4/c1-4-5-6-7-8-9-10-11-12-13-14-17-22(18-15-16-20(24)25)23(26)21(2,3)19-27-22/h4-19H2,1-3H3,(H,24,25)
InChI key
PYDZKXQLRMNQFK-UHFFFAOYSA-N
Application
Lipid spin label used for many types of electron paramagnetic resonance studies including:
- Studies to elucidate the interactions of molecules with the membrane surface
- Kinetics of lipid transfer between lipid bilayers
- Behavior of molecules within a lipid bilayer
- Probing the fluid dynamics of the plasma membrane
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of colloid and interface science, 278(2), 310-317 (2004-09-29)
Interactions and characteristics of 0.1% bovine serum albumin (BSA)-sodium dodecyl sulfate (SDS) in 20 mM phosphate buffer solution were investigated by means of fluorescence spectroscopy and electron spin resonance (ESR) spectroscopy. In BSA-SDS system, the intensity ratio, Im3/Im1, of the
Biochimica et biophysica acta, 1189(2), 233-241 (1994-01-19)
Novel cerebroside sulfate (CBS) spin labels containing long chain C24 or C26 fatty acids with a nitroxide spin label on the 22nd carbon were synthesized and used to investigate the ability of the long fatty acid chains of glycosphingolipids to
Lipids, 45(1), 91-96 (2010-01-06)
Electron paramagnetic resonance (EPR) in conjunction with a slow-tumbling simulation was utilized for defining stratum corneum (SC) lipid structure. We found that ordering calculated from the simulation is an appropriate index for evaluating SC lipids structure. The SC from two
European biophysics journal : EBJ, 39(4), 657-668 (2009-10-14)
Here, we investigate the relationships between the chemical stability of the membrane surface and seed longevity. Dry embryos of long-lived tomato and short-lived onion seeds were labeled with 5-doxyl-stearic acid (5-DS). Temperature-induced loss of the electron spin resonance signal caused
The journal of physical chemistry. B, 115(25), 8054-8062 (2011-05-20)
Insulin entrapment within a monoolein-based reverse hexagonal (H(II)) mesophase was investigated under temperature-dependent conditions at acidic (pH 3) and basic (pH 8) conditions. Studying the structure of the host H(II) system and the interactions of insulin under temperature-dependent conditions has
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service