Skip to Content
Merck
All Photos(3)

Key Documents

216844

Sigma-Aldrich

Magnesium bromide hexahydrate

99%

Synonym(s):

Magnesium dibromide hexahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€81.20
1 KG
€260.00

€81.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 G
€81.20
1 KG
€260.00

About This Item

Linear Formula:
MgBr2 · 6H2O
CAS Number:
Molecular Weight:
292.20
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
crystals and lumps

€81.20


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

crystals and lumps

density

2 g/mL at 25 °C (lit.)

SMILES string

O.O.O.O.O.O.Br[Mg]Br

InChI

1S/2BrH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

LGLXXNHIGIJYQQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium bromide hexahydrate is a white crystalline solid used in the field of catalysis and energy storage devices.[1]

Application

Magnesium bromide hexahydrate can be used:
  • As a catalyst to synthesize dihydropyrimidines via a one-pot Biginelli reaction.[1]
  • As an electrolyte and active bromine source for electrochemica loxidation of sec-alcohols.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Magnesium Bromide and Magnesium Chloride Hexahydrate Catalyzed One-Pot Synthesis of Dihydropyrimidines via Biginelli Reaction under Solvent-Free Conditions
Sirin Gulten
Journal of Heterocyclic Chemistry, 50, 391-395 (2013)
Electrochemical oxidation of sec-alcohols with MgBr2?6H2O
Kosuke Yamamoto, et al.},
Tetrahedron, 1, 100010-100010 (2023)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service