Skip to Content
Merck
All Photos(1)

Key Documents

252263

Sigma-Aldrich

1-Bromo-2-ethylbutane

97%

Synonym(s):

3-(Bromomethyl)pentane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2CHCH2Br
CAS Number:
Molecular Weight:
165.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.4498 (lit.)

bp

143-144 °C (lit.)

density

1.179 g/mL at 25 °C (lit.)

functional group

alkyl halide
bromo

SMILES string

CCC(CC)CBr

InChI

1S/C6H13Br/c1-3-6(4-2)5-7/h6H,3-5H2,1-2H3

InChI key

KKGUMGWNFARLSL-UHFFFAOYSA-N

Application

1-Bromo-2-ethylbutane was used in the preparation of dialkoxytriarylamine[1]. It was also used as cysteine-modifying reagent[2].
Reactant involved in the synthesis of biologically active molecules including:
  • Nantenine analogs with aporphinoid 5-HT2A and α-1A antagonistic activity
  • Aporphines with cytotoxicity toward human colon cancer cell lines
  • Dimeric amide foldamers acting as alpha-helix mimetics
  • Endocrine-disrupting nanylphenol isomers synthesized via demethylation
  • Phosphonic acid-containing benzimidazoles for use as fructose-1,6-biphosphatase inhibitors
  • 5-Bromo-1H-indole-3-acetohydroxamic acid for use as peptide deformylase inhibitors


Reactant involved in studies of the effect of oxygenated side chains in Ephedra compounds

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

96.8 °F - closed cup

Flash Point(C)

36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced poling efficiency in highly thermal and photostable nonlinear optical chromophores.
Faccini M, et al.
Journal of Materials Chemistry, 18(18), 2141-2149 (2008)
Usama M Hegazy et al.
Chemistry & biology, 13(9), 929-936 (2006-09-21)
Proteins contain amino acid residues essential to structure and function. Ribosomal protein synthesis is typically limited to the 20 amino acids of the genetic code, but posttranslational chemical modifications can greatly expand the diversity of side chain functionalities. In this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service