Merck
  • Replacement surgery with unnatural amino acids in the lock-and-key joint of glutathione transferase subunits.
Replacement surgery with unnatural amino acids in the lock-and-key joint of glutathione transferase subunits.
Chemistry & biology (2006-09-21)
Usama M Hegazy, Ulf Hellman, Bengt Mannervik
ABSTRACT
Proteins contain amino acid residues essential to structure and function. Ribosomal protein synthesis is typically limited to the 20 amino acids of the genetic code, but posttranslational chemical modifications can greatly expand the diversity of side chain functionalities. In this investigation, a natural aromatic residue in the lock-and-key joint at the subunit interface of the dimeric glutathione transferase P1-1 was replaced by an S-alkylcysteine residue to give a functional enzyme. Introduction of Cys in the key position inactivates the enzyme, but subsequent alkylation of this residue enhances the catalytic efficiency up to 27,000-fold. Combinatorial modification of Cys by a mixture of reagents facilitated identification of an n-butyl group as the most efficient activator. Alkylation also enhanced binding affinity for active-site ligands and stabilized the enzyme against chemical denaturation and thermal inactivation.
Sigma-Aldrich
(Bromomethyl)cyclobutane, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
1-Iodobutane, 99%, contains copper as stabilizer
Pricing and availability is not currently available.