Skip to Content
Merck
All Photos(1)

Key Documents

250724

Sigma-Aldrich

1,4,8,11-Tetrathiacyclotetradecane

95%

Synonym(s):

14-Ane-S4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20S4
CAS Number:
Molecular Weight:
268.53
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

mp

118-121 °C (lit.)

SMILES string

C1CSCCSCCCSCCSC1

InChI

1S/C10H20S4/c1-3-11-7-9-13-5-2-6-14-10-8-12-4-1/h1-10H2

InChI key

WMVHWZHZRYGGOO-UHFFFAOYSA-N

General description

1,4,8,11-Tetrathiacyclotetradecane is a tetradentate thioether and forms complexes with ruthenium[1]. It is also referred as thio-crown [14]aneS(4) or sulfur crown-ether analog[2].

Application

1,4,8,11-Tetrathiacyclotetradecane has been used in liquid-liquid extraction of metal ions[3].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophie H Dale et al.
Acta crystallographica. Section C, Crystal structure communications, 61(Pt 6), o411-o413 (2005-06-03)
Investigations of the reactivity of the thio-crown [14]aneS(4) (1,4,8,11-tetrathiacyclotetradecane) towards the aminating agent o-mesitylsulfonylhydroxylamine have lead to the crystallization of 1lambda(4),4lambda(4),8lambda(4),11lambda(4)-tetrathiacyclotetradecane-1,4,7,10-tetraylidenetetraaminium tetrakis(2,4,6-trimethylbenzenesulfonate), C10H28N4S(4)4+.4C9H11O3S-. The compound crystallizes in a centrosymmetric space group, with half a formula unit in the asymmetric unit.
Siu-Chung Chan et al.
Inorganic chemistry, 50(22), 11636-11643 (2011-10-26)
Ruthenium complexes containing 2-(2-nitrosoaryl)pyridine (ON(^)N) and tetradentate thioether 1,4,8,11-tetrathiacyclotetradecane ([14]aneS4), [Ru(ON(^)N)([14]aneS4)](2+) [ON(^)N = 2-(2-nitrosophenyl)pyridine (2a), 10-nitrosobenzo[h]quinoline (2b), 2-(2-nitroso-4-methylphenyl)pyridine, (2c), 2-(2-nitrosophenyl)-5-(trifluoromethyl)pyridine (2d)] and analogues with the 1,4,7-trithiacyclononane ([9]aneS3)/tert-butylisocyanide ligand set, [Ru(ON(^)N)([9]aneS3)(C≡N(t)Bu)](2+) (4a and 4b), have been prepared by insertion of a
Liquid-liquid extraction of metal ions by the thiacrown compound 1, 4, 8, 11-tetrathiacyclotetradecane.
Saito K, et al.
Analytica Chimica Acta, 151, 447-455 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service