Skip to Content
Merck
All Photos(1)

Documents

11382

Sigma-Aldrich

1-Aza-18-crown-6

≥98.0% (NT)

Synonym(s):

1,4,7,10,13-Pentaoxa-16-azacyclooctadecane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H25NO5
CAS Number:
Molecular Weight:
263.33
Beilstein:
1074079
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (NT)

form

solid

mp

46-49 °C (lit.)

SMILES string

C1COCCOCCOCCOCCOCCN1

InChI

1S/C12H25NO5/c1-3-14-5-7-16-9-11-18-12-10-17-8-6-15-4-2-13-1/h13H,1-12H2

InChI key

NBXKUSNBCPPKRA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Multicomponent pseudorotaxane quadrilateral as dual-way logic AND gate with two catalytic outputs: This study explores the use of 1-Aza-18-crown-6 in a multicomponent system functioning as a dual-way logic AND gate, demonstrating its potential in molecular electronics and sensing (Kundu et al., 2022).
  • Facile fabrication of crown ether functionalized lignin-based biosorbent for the selective removal of Pb (II): This research developed a 1-Aza-18-crown-6 functionalized lignin for effective lead removal from water, highlighting its utility in environmental cleanup (Jin et al., 2020).
  • Synthesis of helical polyisocyanides bearing aza‐crown ether groups as pendant: This study involves the synthesis of helical polymers with pendant aza-crown ether groups, opening new avenues for the development of advanced materials (Kanbayashi et al., 2018).
  • 1-aza-18-crown-6 ether tailored graphene oxide for Cs (I) removal from wastewater: Demonstrates the modification of graphene oxide with 1-Aza-18-crown-6 for cesium ion removal from wastewater, an important application in radioactive waste management (Liu et al., 2023).

Other Notes

Building block for the synthesis of bis-crown ethers, lariat ethers and other monofunctionalized macrocycles

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G.W. Gokel and J.E. Trafton et al.
Cation Binding by Macrocycles null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service