Skip to Content
Merck
All Photos(1)

Key Documents

244937

Sigma-Aldrich

Dichloro(1,5-cyclooctadiene)platinum(II)

Dichloro(1,5-cyclooctadiene)platinum(II)

Synonym(s):

(1,5-Cyclooctadiene)platinum(II) dichloride, 1,5-Cyclooctadienedichloroplatinum, (1,5-Cyclooctadiene)platinum dichloride, NSC 239486, Pt(COD)Cl2

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€45.00
1 G
€171.00

€45.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€45.00
1 G
€171.00

About This Item

Empirical Formula (Hill Notation):
C8H12Cl2Pt
CAS Number:
Molecular Weight:
374.16
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€45.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

reaction suitability

core: platinum
reagent type: catalyst

mp

285 °C (dec.) (lit.)

SMILES string

Cl[Pt]Cl.C1CC=CCCC=C1

InChI

1S/C8H12.2ClH.Pt/c1-2-4-6-8-7-5-3-1;;;/h1-2,7-8H,3-6H2;2*1H;/q;;;+2/p-2/b2-1-,8-7-;;;

InChI key

VVAOPCKKNIUEEU-PHFPKPIQSA-L

Application

Catalyst for:
  • Hydrogenative cyclization of allenynes
  • Stereoselective tandem hydrosilylation-Hiyama coupling reactions
  • Amino alkene cyclohydroamination
  • Hydrative cyclization reactions
  • Direct amination of allylic alcohols under mild conditions

  • Potential anticancer agent

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiaojiao Xu et al.
Soft matter, 15(30), 6116-6126 (2019-07-10)
In this work, according to the characteristic of surface plasmon resonance (SPR) of metallic nanoparticles, we investigated the photo actuation performance of a liquid crystalline elastomer (LCE) nanocomposite with incorporated gold nanoparticles (nano-gold/LCE nanocomposite). The nano-gold/LCE nanocomposites were fabricated by
Deborah Vidick et al.
Beilstein journal of nanotechnology, 6, 1287-1297 (2015-07-23)
Heterometal clusters containing Ru and Au, Co and/or Pt are anchored onto carbon nanotubes and nanofibers functionalized with chelating phosphine groups. The cluster anchoring yield is related to the amount of phosphine groups available on the nanocarbon surface. The ligands

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service