Skip to Content
Merck
All Photos(1)

Key Documents

234532

Sigma-Aldrich

Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid

97%

Synonym(s):

EGTA, Egtazic acid, Ethylene-bis(oxyethylenenitrilo)tetraacetic acid, Glycol ether diamine tetraacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2OCH2CH2N(CH2CO2H)2]2
CAS Number:
Molecular Weight:
380.35
Beilstein:
1717370
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

97%

mp

241 °C (dec.) (lit.)

SMILES string

OC(=O)CN(CCOCCOCCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C14H24N2O10/c17-11(18)7-15(8-12(19)20)1-3-25-5-6-26-4-2-16(9-13(21)22)10-14(23)24/h1-10H2,(H,17,18)(H,19,20)(H,21,22)(H,23,24)

InChI key

DEFVIWRASFVYLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A chelating agent useful for the determination of calcium in the presence of magnesium.

Biochem/physiol Actions

Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid A is a chelating agent used for the determination of calcium in the presence of magnesium.

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeison García et al.
Journal of medicinal chemistry, 53(2), 811-821 (2009-12-17)
Two widely studied parasite protein families are considered attractive targets for developing a fully effective antimalarial vaccine: the erythrocyte binding antigen (EBA) family defining a sialic acid-dependent invasion pathway, and reticulocyte-binding homologue (RH) proteins associated with sialic acid-independent red blood
Yesid Garcia et al.
Journal of medicinal chemistry, 50(23), 5665-5675 (2007-10-20)
Plasmodium falciparum merozoite surface proteins (MSP-1 to -11) have been involved in merozoite interaction with the red blood cell (RBC) surface. Peptides covering complete MSP-4 and MSP-7 amino acid sequences were synthesized and tested in RBC binding assays. One MSP-4
Valery N Danilenko et al.
Journal of medicinal chemistry, 51(24), 7731-7736 (2008-12-05)
Aminomethylation of 9b,10-dihydro-1H-indolo[1,7:4,5,6]pyrrolo[3,4:2,3][1,4]diazepino-[1,7-a]indole-1,3(2H)-diones or 1H-indolo[1,7:4,5,6]pyrrolo[3,4:2,3][1,4]diazepino[1,7-a]indole-1,3(2H)-diones resulted in dialkylaminomethyl derivatives. Alkylation of the nitrogen atom of maleimide moiety of polyannelated diazepines with 1,3-dibromopropane and subsequent reaction with thiourea or its N-alkyl derivatives gave isothiourea-carrying compounds. The compounds containing isothiourea moiety were
Ana Zuleima Obando-Martinez et al.
Journal of medicinal chemistry, 53(10), 3907-3918 (2010-04-24)
Detergent resistant membranes (DRMs) of Plasmodium falciparum merozoites contain a large number of glycosylphosphatidylinositol (GPI)-anchored proteins that have been implicated in interactions between merozoites and red blood cells (RBCs). In this study, two cysteine-rich proteins anchored by GPI to merozoite
Phedias Diamandis et al.
Nature chemical biology, 3(5), 268-273 (2007-04-10)
The identification of self-renewing and multipotent neural stem cells (NSCs) in the mammalian brain holds promise for the treatment of neurological diseases and has yielded new insight into brain cancer. However, the complete repertoire of signaling pathways that governs the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service