Merck
  • Search for inhibitors of bacterial and human protein kinases among derivatives of diazepines[1,4] annelated with maleimide and indole cycles.
Search for inhibitors of bacterial and human protein kinases among derivatives of diazepines[1,4] annelated with maleimide and indole cycles.
Journal of medicinal chemistry (2008-12-05)
Valery N Danilenko, Alexander Y Simonov, Sergey A Lakatosh, Michael H G Kubbutat, Frank Totzke, Christoph Schächtele, Sergey M Elizarov, Olga B Bekker, Svetlana S Printsevskaya, Yuryi N Luzikov, Marina I Reznikova, Alexander A Shtil, Maria N Preobrazhenskaya
ABSTRACT
Aminomethylation of 9b,10-dihydro-1H-indolo[1,7:4,5,6]pyrrolo[3,4:2,3][1,4]diazepino-[1,7-a]indole-1,3(2H)-diones or 1H-indolo[1,7:4,5,6]pyrrolo[3,4:2,3][1,4]diazepino[1,7-a]indole-1,3(2H)-diones resulted in dialkylaminomethyl derivatives. Alkylation of the nitrogen atom of maleimide moiety of polyannelated diazepines with 1,3-dibromopropane and subsequent reaction with thiourea or its N-alkyl derivatives gave isothiourea-carrying compounds. The compounds containing isothiourea moiety were active against individual human serine/threonine and tyrosine kinases at low micromolar concentrations. Dialkylaminomethyl derivatives of diazepines sensitized Streptomyces lividans with overexpressed aminoglycoside phosphotransferase type VIII (aphVIII) to kanamycin by inhibiting serine/threonine kinase(s) mediated aphVIII phosphorylation.
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
KN-62, ≥95%, powder