Skip to Content
Merck
All Photos(1)

Key Documents

SYNP170004D

Synthware round bottom pressure vessel with protective coating

capacity 150 mL, O.D. × L 60 mm × 100 mm, bushing type, #25

Synonym(s):

Synthware P170004D, pressure tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

feature

bushing type #25

packaging

pack of 1 ea

manufacturer/tradename

Kemtech P170004D

O.D. × L

60 mm × 100 mm

capacity

150 mL

Looking for similar products? Visit Product Comparison Guide

General description

Pressure Vessel, Heavy Wall, Rb, 150mL, #25, 60X100mm, Protective Covering, Working Temp -40A°C To 80A°C,Includes Accessories: B100025 + R212EP

Application

kemetch; synthware; pressure vessel; cylindrical

Features and Benefits

Organic Chemistry

Legal Information

Synthware is a trademark of Kemtech America Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service