Skip to Content
Merck
All Photos(1)

Key Documents

SYNP170003D

Synthware round bottom pressure vessel with protective coating

bottle capacity 150 mL, O.D. × L 60 mm × 100 mm, bushing type, #15

Synonym(s):

Synthware P170003D, pressure tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

feature

bushing type #15

packaging

pack of 1 ea

manufacturer/tradename

Kemtech P170003D

O.D. × L

60 mm × 100 mm

bottle capacity

150 mL

General description

Pressure Vessel, Heavy Wall, Rb, 150mL, #15, 60X100mm, Protective Covering, Working Temp -40°C To 80°C,Includes Accessories: B100015 + R111Ep

Application

kemetch; synthware; pressure vessel; round bottom

Features and Benefits

Organic Chemistry

Legal Information

Synthware is a trademark of Kemtech America Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service