Skip to Content
Merck
All Photos(1)

Key Documents

SML3594

Sigma-Aldrich

Masitinib

≥98% (HPLC)

Synonym(s):

4-[(4-Methyl-1-piperazinyl)methyl]-N-[4-methyl-3-[[4-(3-pyridinyl)-2-thiazolyl]amino]phenyl]-benzamide, AB1010

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H30N6OS
CAS Number:
Molecular Weight:
498.64
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=CC=C(NC(C2=CC=C(CN3CCN(C)CC3)C=C2)=O)C=C1NC4=NC(C5=CN=CC=C5)=CS4

InChI

1S/C28H30N6OS/c1-20-5-10-24(16-25(20)31-28-32-26(19-36-28)23-4-3-11-29-17-23)30-27(35)22-8-6-21(7-9-22)18-34-14-12-33(2)13-15-34/h3-11,16-17,19H,12-15,18H2,1-2H3,(H,30,35)(H,31,32)

InChI key

WJEOLQLKVOPQFV-UHFFFAOYSA-N

Biochem/physiol Actions

Masitinib is a protein tyrosine kinase inhibitor that selectively targets stem cell factor receptor c-kit in particular and also PDGFRalpha/beta and Lyn. It is more potent and selective for c-kit than imatinib (Gleevec). Masitinib has antineoplastic and anti-inflammatory activity. It inhibits the survival, migration and activity of mast cells, key components of the inflammation response. It is used to treat mast cell tumors in dogs and is in clinical trials for human use in rheumatoid arthritis, asthma, and as an adjuct therapy for Alzheimer′s disease.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masitinib, a c-kit/PDGF receptor tyrosine kinase inhibitor, improves disease control in severe corticosteroid-dependent asthmatics
M Humbert
Allergy, 64, 1194-1201 (2009)
Patrice Dubreuil et al.
PloS one, 4(9), e7258-e7258 (2009-10-01)
The stem cell factor receptor, KIT, is a target for the treatment of cancer, mastocytosis, and inflammatory diseases. Here, we characterise the in vitro and in vivo profiles of masitinib (AB1010), a novel phenylaminothiazole-type tyrosine kinase inhibitor that targets KIT.
Masitinib as an adjunct therapy for mild-to-moderate Alzheimer's disease: a randomised, placebo-controlled phase 2 trial
Francois Piette
Alzheimer's Research & Therapy, 3, 16-16 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service