Skip to Content
Merck
All Photos(1)

Documents

SML2248

Sigma-Aldrich

SK-216

≥98% (HPLC)

Synonym(s):

2-[5-[[6-[5-(1,1-Dimethylethyl)-2-benzoxazolyl]-2-naphthalenyl]oxy]pentyl]-propanedioic acid sodium salt, SK 216, [5-[[6-[5-(1,1-dimethylethyl)-2-benzoxazolyl]-2-naphthalenyl]oxy]pentyl]-propanedioic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H29NNa2O6
CAS Number:
Molecular Weight:
533.52
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 0.5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

CC(C)(C)C(C=C1)=CC2=C1OC(C3=CC=C4C(C=CC(OCCCCCC(C([O-])=O)C([O-])=O)=C4)=C3)=N2.[2Na+]

InChI

1S/C29H31NO6.2Na/c1-29(2,3)21-11-13-25-24(17-21)30-26(36-25)20-9-8-19-16-22(12-10-18(19)15-20)35-14-6-4-5-7-23(27(31)32)28(33)34;;/h8-13,15-17,23H,4-7,14H2,1-3H3,(H,31,32)(H,33,34);;/q;2*+1/p-2

InChI key

YYTGMMYYLGKWIK-UHFFFAOYSA-L

Biochem/physiol Actions

SK-216 is an orally active potent and selective inhibitor of Plasminogen activator inhibitor-1 (PAI-1) that inhibits tumor progression and angiogenesis. SK-216 inhibits migration and tube formation of cultured human umbilical vein endothelial cells induced by various angiogenic factors. SK-116 prevents a formation of gastro-intestinal polyps in Min mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service