Skip to Content
Merck
All Photos(1)

Key Documents

SML1679

Sigma-Aldrich

DEL-22379

≥98% (HPLC)

Synonym(s):

N-(3-((5-Methoxy-1H-indol-3-yl)methylene)-2-oxoindolin-5-yl)-3-(piperidin-1-yl)propanamide, N-[2,3-Dihydro-3-[(5-methoxy-1H-indol-3-yl)methylene]-2-oxo-1H-indol-5-yl]-1-piperidinepropanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H28N4O3
CAS Number:
Molecular Weight:
444.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C1NC2=CC=C(NC(CCN3CCCCC3)=O)C=C2C1=CC4=CNC5=C4C=C(OC)C=C5

InChI key

INQUULPXCZAKMS-UHFFFAOYSA-N

Biochem/physiol Actions

DEL-22379 is a cell-permeable 3-arylidene-2-oxindole derivative that inhibits the dimerization of ERK (IC50 = 500 nM) via directly binding to its dimerization interface. DEL-22379 promotes the transactivation of ELK, and prevents the EGF-induced phosphorylation of the cytoplasmic kinase RSK1 in HEK293 cells (IC50 = 500 nM). DEL-22379 induces apoptosis in various cancer cells harboring RAS-ERK pathway oncogenes (IC50 = 150 – 400 nM). DEL-22379 blocks tumor progression and metastasis in multiple xenograft models such as A375 (BRAF mutant), HCT116 (KRAS mutant) and colorectal cancer patient-derived xenografts (15 mg/kg, i.p. b.i.d.).
DEL-22379 is a cell-permeable 3-arylidene-2-oxindole derivative that inhibits the dimerization of ERK via directly binding to its dimerization interface.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service