All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C46H64N14O12S2
Molecular Weight:
1069.22
UNSPSC Code:
12352209
NACRES:
NA.32
Recommended Products
Assay
≥95% (HPLC)
form
lyophilized
composition
Peptide Content, ≥75%
storage condition
protect from light
storage temp.
−20°C
Related Categories
Amino Acid Sequence
Mpa-Tyr-Phe-D-Gln-Asn-Cys-Pro-D-Arg-Gly-NH2
Application
Desmopressin is a synthetic alternative to arginine vasopressin that acts via V2 receptors in renal collecting ducts to regulate water retention. [D-Gln4]-Desmopressin may be used in studies of conformations and receptor activity of desmopressin analogues.
Biochem/physiol Actions
Desmopressin is considered safe for consumption during first and second trimesters of pregnancy. But, it shows high risk of arterial thrombosis. It is known to induce platelet factor VIII and VWF (Von Willebrand factor) and improves platelet aggregation. Desmopressin is found helpful in congenital bleeding disorder.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Hemostasis and Thrombosis: Basic Principles and Clinical Practice, 63-63 (2014)
Platelets, 1243-1243 (2011)
Mattias Hedenström et al.
Journal of medicinal chemistry, 45(12), 2501-2511 (2002-05-31)
Three analogues of the antidiuretic drug desmopressin ([1-desamino,8-D-arginine]vasopressin) have been prepared. In two of these, gamma-turn mimetics based on a morpholin-3-one framework have been inserted instead of residues Phe3-Asn5, whereas the third analogue has a methylene ether isostere in place
J Wang et al.
International journal of peptide and protein research, 45(5), 471-481 (1995-05-01)
The solution structures of desmopressin, a nine-residue peptide with specific antidiuretic and antibleeding activities, have been studied in aqueous and TFE-containing solutions by two-dimensional NMR and molecular modeling techniques. It is found that TFE induces a conformational change of the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service