Skip to Content
Merck
All Photos(1)

Key Documents

S4647

Sigma-Aldrich

Glass spreaders

120° bend

Synonym(s):

cell spreader, glass spreader

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122103
NACRES:
NB.14

General description

Constructed of 4 mm diameter glass rod with polished ends, with 130 mm long handles and 50 mm long spreader segments.

Application

For inoculating petri dishes

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service