Skip to Content
Merck
All Photos(2)

Documents

90922

Millipore

Mueller Hinton Broth 2

NutriSelect® Plus, suitable for microbiology, Cation-adjusted

Synonym(s):

M-H 2 Broth, M-H 2 Broth, Mueller Hinton II Broth, Mueller Hinton II Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171504
NACRES:
NA.85

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

acid hydrolysate of casein, 17.5 g/L
beef extract, 3 g/L
starch, 1.5 g/L

packaging

bottle of 500 g

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | susceptibility testing: suitable
microbiological culture: suitable

color

light yellow

final pH

7.3±0.2 (25 °C)

application(s)

clinical testing
food and beverages

microbiology

suitability

nonselective for Enterococcus spp.
nonselective for Escherichia coli
nonselective for Neisseria spp.
nonselective for Pseudomonas spp.
nonselective for Staphylococcus spp.
nonselective for bacteria (General Media)
nonselective for coliforms

General description

Mueller Hinton Broth 2 is recommended for testing the most encountered aerobic and facultative anaerobic bacteria found in food and clinical material for antimicrobial susceptibility testing. The medium shows good batch-to-batch reproducibility. It is low in sulfonamide, trimethoprim, and tetracycline inhibitors and yields satisfactory growth of most non-fastidious pathogens.Casein acid hydrolysate and beef extract provide amino acids and other nitrogenous substances, minerals, vitamins, and carbon to support the growth of microorganisms. To determine the MIC values with Enterococcus faecalis and sulfamethoxazole-trimethoprim, a special low content of thymine and thymidine in the amino acid sources is required. Starch neutralizes toxic substances that may be present in the medium and have an inhibitory effect on antibiotics and the growth of organisms. During autoclaving, a small amount of dextrose is hydrolyzed from starch, which is a source of energy. Calcium and magnesium salts are added to adjust the ion concentrations of the medium.

Application

Mueller Hinton Broth 2 cation adjusted is intended for use in quantitative procedures for susceptibility testing of rapidly growing aerobic and facultatively anaerobic bacteria isolated from clinical specimens. It has been used for culturing Escherichia coli, Bacillus subtilis, Staphylococcus aureus and in the serial dilution of doxycycline.

Features and Benefits

NutriSelect® Plus are powdered culture media offering
  • Quality control with growth promotion test according to industry standards
  • Quality assurance acc. to ISO 9001
  • Cost-efficient powdered media

Preparation Note

Suspend 22 g in 1000 ml distilled water. Heat if necessary to dissolve the medium completely. Dispense and sterilize by autoclaving at 10-15 lbs pressure (115-121°C) for 10 minutes. DO NOT OVERHEAT. Mix well before pouring.
Note: This medium is supplemented with appropriate salts to provide 20-25 mg/l of calcium and 10-12.5 mg/l of magnesium and as additionally required to suit performance parameters.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joshua S Moskowitz et al.
Biomaterials, 31(23), 6019-6030 (2010-05-22)
While the infection rate of orthopedic implants is low, the required treatment, which can involve six weeks of antibiotic therapy and two additional surgical operations, is life threatening and expensive, and thus motivates the development of a one-stage re-implantation procedure.
Inês Martins-Oliveira et al.
European journal of clinical microbiology & infectious diseases : official publication of the European Society of Clinical Microbiology, 39(10), 1907-1914 (2020-06-03)
The urgent need for rapid antimicrobial susceptibility is broadly apparent from government reports to the lay press. Accordingly, we developed a flow-cytometry assay (FCM) for evaluating ceftolozane-tazobactam (C/T) susceptibility directly on blood cultures (BC) requiring < 2 h from flag
Mohaned Shilaih et al.
Antiviral therapy, 23(1), 91-94 (2017-05-13)
Despite being used by more than 18 million people our understanding of the extent of the effects of antiretrovirals on the human body and other organisms remains incomplete. In addition, the direct effect of antiretrovirals on the gut microbiota of
Thomas Kohler et al.
Journal of bacteriology, 191(13), 4482-4484 (2009-05-12)
Skin-colonizing gram-positive bacteria produce wall teichoic acids (WTAs) or related glycopolymers for unclear reasons. Using a WTA-deficient Staphylococcus aureus mutant, we demonstrated that WTA confers resistance to antimicrobial fatty acids from human sebaceous glands by preventing fatty acid binding. Thus
Elizabeth I Parkinson et al.
Nature chemical biology, 15(11), 1049-1056 (2019-08-28)
Fosmidomycin and related molecules comprise a family of phosphonate natural products with potent antibacterial, antimalarial and herbicidal activities. To understand the biosynthesis of these compounds, we characterized the fosmidomycin producer, Streptomyces lavendulae, using biochemical and genetic approaches. We were unable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service