Skip to Content
Merck
All Photos(1)

Key Documents

GE17-3717-55

HiTrap® Capto S ImpAct 5x5mL

Cytiva 17-3717-55, pack of 5x5ml

Synonym(s):

HITRAP® CAPTO S IMPACT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.56
Pricing and availability is not currently available.

ligand

(-SO3)

manufacturer/tradename

Cytiva 17-3717-55

matrix

high-flow agarose

avg. part. size

50 μm (d50V)

cleaning in place

4-12

working range

3-14

capacity

binding capacity(>85 mg BSA/mL medium, >90 mg lysozyme/ml medium, >100 mg IgG/ ml medium)

Looking for similar products? Visit Product Comparison Guide

General description

HiTrap Capto S ImpAct (1 ml and 5 ml) are prepacked columns for small scale purifications, as well as optimization of methods and parameters, such as sample load and binding conditions. HiTrap Capto S ImpAct columns provide fast, reproducible and easy separations in a convenient format. The columns are best utilized when they are connected to liquid chromatography systems such as ÄKTA systems.

Capto S ImpAct is a BioProcess chromatography medium developed for large-scale purification of biopharmaceutical proteins. Based on the well-established Capto base matrix, it provides many opportunities for improved productivity and straightforward process development. This chromatography medium (resin) is available in a range of different bulk pack sizes and convenient prepacked formats for easy scale-up and process development. As a member of the BioProcess media range, Capto S ImpAct meets industrial demands with security of supply and comprehensive technical and regulatory support.

Features and Benefits

HiTrap Capto S ImpAct columns are prepacked with Capto S ImpAct cation exchange chromatography medium for screening and small-scale protein purifications using ion exchange chromatography (IEX).

  • Convenient prepacked column for fast, reproducible, and easy small-scale purifications.
  • High binding capacity, > 100 mg MAb/mL medium.
  • Efficient aggregate removal at high load of monoclonal antibodies.
  • High-resolution intermediate purification and polishing based on the well-established Capto platform.
  • Flexibility of design thanks to a large operational window of flow rates and bed heights for easy optimization and scaling.

Storage and Stability

Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Capto is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service