GE28-4010-81
HiTrap® _HiPrep™ 1/16 in. Male Connector For Akta Design
Cytiva 28-4010-81, pack of 8 pieces
All Photos(1)
About This Item
UNSPSC Code:
41115711
NACRES:
NA.56
Recommended Products
material
polypropylene
feature
wetted part
packaging
pack of 8 pieces
manufacturer/tradename
Cytiva 28-4010-81
General description
Connector 1/16" male to use with ÄKTA start, ÄKTA go, ÄKTA pure, and ÄKTA avant.
Features and Benefits
- Fingertight
- Narrow
- For connecting HiTrap and HiPrep columns
Legal Information
HiPrep is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva
related product
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)