Skip to Content
Merck
All Photos(1)

Key Documents

CLC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

fish blood

packaging

tube of 5 μg 95070628-DNA-5UG
pkg of vial of cells 95070628-1VL

growth mode

Adherent

karyotype

2n = 104

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Carp leukocyte

Cell Line Description

The cell line CLC (Carp Leukocyte Culture) was established from peripheral blood mononuclear cells of a normal 2 year old female common carp, Cyprinus carpio. They possess phagocytic activity and are reported to be stimulated by TNF, gamma-interferon and LPS therefore resembling characteristic features of monocytes / macrophages. CLC replicate fish viruses e.g. IPNV, PFR, SVCV and VHSV.

Application

Immunological research, fish virology, fish cell physiology.

Culture Medium

EMEM (EBSS) or TC-199 + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Spilt confluent cultures 1:3 using 0.25% trypsin or trypsin/EDTA; 25-28°C. Fish cell lines detach easily during transit if the culture is too young. After resuscitation split cells 1:3 and culture for at least one week with 1-2 media changes before shipping.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service