Skip to Content
Merck
All Photos(1)

Key Documents

Grunt Fin (GF)

88010601, fish fin, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

Grunt Fin (GF), 88010601

biological source

fish fin

growth mode

Adherent

karyotype

Modal no. 47

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Fish Grunt fin tissue blue striped

Cell Line Description

Derived from fin tissue of the adult salt water blue striped Grunt. Does not support the replication of influenza A, herpes simplex, adeno, polio, dengue and eastern equine encephalitis viruses.

Culture Medium

EMEM (HBSS) (Sigma cat no. M5775) + 10mM Hepes (pH 7.3) + 2mM Glutamine + 0.23% (w/v) NaHCO3 + 20% FBS (Heat Inactivated)+ 58mM NaCl

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 20°C. After resuscitation split cells 1:2 to 1:4 and culture for at least one week with 1-2 media changes before shipping. Plating efficiency can be very low a

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service