902292
EvanPhos
≥95%
Synonym(s):
Biaryl phosphine ligand for Pd catalysis, Dicyclohexyl(2,6-dimethoxy-3-(2-methoxynaphthalen-1-yl)phenyl)phosphane
About This Item
Recommended Products
Assay
≥95%
form
powder or crystals
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Cross Couplings
greener alternative product characteristics
Catalysis
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
mp
131-133 °C
functional group
phosphine
greener alternative category
SMILES string
P(C5CCCCC5)(C4CCCCC4)c1c(c(ccc1OC)c2c3c(ccc2OC)cccc3)OC
InChI key
IJZUTUMBEYLTNN-UHFFFAOYSA-N
General description
Application
Other Notes
Watch Professor Lipshutz talk about greener chemistry in this webinar: From milligrams to kilograms: synthetic chemistry following nature′s lead
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service