Skip to Content
Merck
All Photos(1)

Key Documents

86837

Supelco

Tetrabutylammonium benzoate

for electrochemical analysis, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(OCOC6H5)
CAS Number:
Molecular Weight:
363.58
Beilstein:
4070140
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

Quality Level

Assay

≥99.0% (NT)
≥99.0%

form

crystals

mp

64-67 °C (lit.)

solubility

acetonitrile: 0.1 g/mL, clear, colorless

SMILES string

[O-]C(=O)c1ccccc1.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.C7H6O2/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;8-7(9)6-4-2-1-3-5-6/h5-16H2,1-4H3;1-5H,(H,8,9)/q+1;/p-1

InChI key

WGYONVRJGWHMKV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Tetrabutylammonium benzoate may be used as an analytical reagent for the electrochemical generation of hydrogen from acetic acid using a molecular molybdenum-oxo catalyst.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical generation of hydrogen from acetic acid using a molecular molybdenum?oxo catalyst.
Thoi SV, et al.
Energy & Environmental Science, 5(7), 7762-7770 (2012)
Daniel J Martin et al.
Science advances, 6(11), eaaz3318-eaaz3318 (2020-03-24)
The development of advanced chemical-to-electrical energy conversions requires fast and efficient electrocatalysis of multielectron/multiproton reactions, such as the oxygen reduction reaction (ORR). Using molecular catalysts, correlations between the reaction rate and energy efficiency have recently been identified. Improved catalysis requires
Ken T Ngo et al.
Journal of the American Chemical Society, 139(7), 2604-2618 (2017-01-25)
Electrocatalytic reduction of CO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service