Skip to Content
Merck
All Photos(1)

Key Documents

8.03541

Sigma-Aldrich

Dichloroacetic acid

for synthesis

Synonym(s):

Dichloroacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H2Cl2O2
CAS Number:
Molecular Weight:
128.94
MDL number:
UNSPSC Code:
12352106
EC Index Number:
201-207-0
NACRES:
NA.22

vapor pressure

0.19 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

2820 mg/kg LD50, oral (Rat)

pH

1.2 (20 °C, 129 g/L in H2O)

bp

192 °C/1013 hPa (decomposes)

mp

-4 °C

density

1.57 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClC(Cl)C(=O)O

InChI

1S/C2H2Cl2O2/c3-1(4)2(5)6/h1H,(H,5,6)

InChI key

JXTHNDFMNIQAHM-UHFFFAOYSA-N

Application

  • Controlled drug release systems: Research includes the application of Dichloroacetic acid in the synthesis of sulfonated polyether ketone membranes that are embedded with nalidixic acid, highlighting its utility in developing emerging controlled drug releasers in medical applications (Padinjarathil et al., 2023).

Linkage

Replaces: DX0653

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.560 - 1.570
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Lact. - Met. Corr. 1 - Repr. 1B - Skin Corr. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service